CH6371377515 Chem Impex International, Inc.

Z-L-threonine a-benzyl ester

  • MDL number MFCD00077041
  • Cas Number : 16597-50-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 77.0-84.0
Synonym Z-L-Thr-OBzl
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-l-threonine-alpha-benzyl-ester/1470?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-l-threonine-alpha-benzyl-ester/1470?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart