CH6371377525 Chem Impex International, Inc.

Z-L-tyrosine 4-nitrophenyl ester

  • MDL number MFCD00024659
  • Cas Number : 3556-56-7
  • Molecular Weight : 436.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8°C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-l-tyrosine-4-nitrophenyl-ester/1480?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-l-tyrosine-4-nitrophenyl-ester/1480?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart