CH6371377527 Chem Impex International, Inc.

Z-D-valine

  • MDL number MFCD00065703
  • Cas Number : 1685-33-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 53-59� C
Synonym Z-D-Val-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-d-valine/1482?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-d-valine/1482?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart