CH6371377596 Chem Impex International, Inc.

Fmoc-L-alanine N-hydroxysuccinimide ester

  • MDL number MFCD00065613
  • Cas Number : 73724-40-0
  • Molecular Weight : 408.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158-164 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-l-alanine-i-n-i-hydroxysuccinimide-ester/1554?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-l-alanine-i-n-i-hydroxysuccinimide-ester/1554?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart