CH6371377603 Chem Impex International, Inc.

Na-Fmoc-Nw-(2,2,5,7,8-pentamethylchroman-6-sulfonyl)-D-arginine

  • MDL number MFCD00065619
  • Cas Number : 157774-30-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-D-Arg(Pmc)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-fmoc-i-n-i-sup-sup-2-2-5-7-8-pentamethylchroman-6-sulfonyl-d-arginine/1562?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-fmoc-i-n-i-sup-sup-2-2-5-7-8-pentamethylchroman-6-sulfonyl-d-arginine/1562?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart