CH6371377605 Chem Impex International, Inc.

Na-Fmoc-Nw-(4-toluenesulfonyl)-D-arginine

  • MDL number MFCD00065621
  • Cas Number : 139090-50-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-D-Arg(Tos)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-fmoc-i-n-i-sup-omega-sup-4-toluenesulfonyl-d-arginine/1564?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-fmoc-i-n-i-sup-omega-sup-4-toluenesulfonyl-d-arginine/1564?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart