CH6371377615 Chem Impex International, Inc.

Fmoc-S-4-methoxybenzyl-L-cysteine

  • MDL number MFCD00065635
  • Cas Number : 141892-41-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138-142
Synonym Fmoc-L-Cys(pMeOBzl)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-s-i-4-methoxybenzyl-l-cysteine/1578?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-s-i-4-methoxybenzyl-l-cysteine/1578?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart