CH6371377622 Chem Impex International, Inc.

Na-Fmoc-Nd-4,4'-dimethoxybenzhydryl-D-glutamine

  • MDL number MFCD00065645
  • Cas Number : 210645-01-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160-180
Synonym Fmoc-Ndelta-4,4'-dimethoxybenzhydryl-D-glutamine
Purity 98
Storage temp. Store at RT
Physical Form White to off white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-fmoc-i-n-i-sup-d-sup-4-4-dimethoxybenzhydryl-d-glutamine/1587?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-fmoc-i-n-i-sup-d-sup-4-4-dimethoxybenzhydryl-d-glutamine/1587?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart