CH6371377631 Chem Impex International, Inc.

Fmoc-D-alpha-tert-butylglycine

  • MDL number MFCD00065650
  • Cas Number : 198543-64-5
  • Molecular Weight : 353.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 136 - 138 °C
Purity 99.5
Storage temp. Store at 0 - 8 °C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-d-a-i-tert-i-butylglycine/1596?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-d-a-i-tert-i-butylglycine/1596?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart