CH6371377637 Chem Impex International, Inc.

Fmoc-L-leucine N-hydroxysuccinimide ester

  • MDL number MFCD00065659
  • Cas Number : 76542-83-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120-125 °C
Purity 98
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-l-leucine-i-n-i-hydroxysuccinimide-ester/1605?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-l-leucine-i-n-i-hydroxysuccinimide-ester/1605?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart