CH6371377638 Chem Impex International, Inc.

Na-Fmoc-Ne-acetyl-L-lysine

  • MDL number MFCD00077409
  • Cas Number : 159766-56-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155-165
Synonym Fmoc-L-Lys(Ac)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-fmoc-i-n-i-sup-epsilon-sup-acetyl-l-lysine/1606?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-fmoc-i-n-i-sup-epsilon-sup-acetyl-l-lysine/1606?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart