CH6371377645 Chem Impex International, Inc.

Fmoc-L-methionine N-hydroxysuccinimide ester

  • MDL number MFCD00065666
  • Cas Number : 112913-64-1
  • Molecular Weight : 468.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129-139 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-l-methionine-i-n-i-hydroxysuccinimide-ester/1615?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-l-methionine-i-n-i-hydroxysuccinimide-ester/1615?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart