CH6371377654 Chem Impex International, Inc.

Fmoc-L-proline N-hydroxysuccinimide ester

  • MDL number MFCD00065673
  • Cas Number : 109074-94-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 161-166 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-l-proline-i-n-i-hydroxysuccinimide-ester/1626?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-l-proline-i-n-i-hydroxysuccinimide-ester/1626?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart