CH6371377659 Chem Impex International, Inc.

Fmoc-(3S,4S)-4-amino-3-hydroxy-6-methylheptanoic acid

  • MDL number MFCD00065675
  • Cas Number : 158257-40-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-Sta(3S,4S)-OH
Purity 99
Storage temp. Store at = -4
Physical Form White powder or white solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-3-i-s-i-4-i-s-i-4-amino-3-hydroxy-6-methylheptanoic-acid/1631?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-3-i-s-i-4-i-s-i-4-amino-3-hydroxy-6-methylheptanoic-acid/1631?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart