CH6371377671 Chem Impex International, Inc.

Fmoc-O-2,6-dichlorobenzyl-D-tyrosine

  • MDL number MFCD00274143
  • Cas Number : 419573-91-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 168 - 169 °C
Purity 99
Storage temp. Store at 0 - 8 °C
Physical Form White crystals
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-o-i-2-6-dichlorobenzyl-d-tyrosine/1644?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-o-i-2-6-dichlorobenzyl-d-tyrosine/1644?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart