CH6371377697 Chem Impex International, Inc.

Boc-D-a-aminobutyric acid dicyclohexylammonium salt

  • MDL number MFCD00235886
  • Cas Number : 27494-47-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-D-Abu-OH DCHA
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-d-a-aminobutyric-acid-dicyclohexylammonium-salt/1708?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-d-a-aminobutyric-acid-dicyclohexylammonium-salt/1708?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart