CH6371377700 Chem Impex International, Inc.

L-2-Cyclohexylglycine hydrochloride

  • MDL number MFCD00050539
  • Cas Number : 14328-51-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Chg-OH HCl
Purity 99
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-2-cyclohexylglycine-hydrochloride/1711?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-2-cyclohexylglycine-hydrochloride/1711?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart