CH6371377720 Chem Impex International, Inc.

Boc-3-(1-naphthyl)-L-alanine

  • MDL number MFCD00085377
  • Cas Number : 55447-00-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-L-Ala(1-naphthyl)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-3-1-naphthyl-l-alanine/1731?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-3-1-naphthyl-l-alanine/1731?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart