CH6371377731 Chem Impex International, Inc.

Boc-S-4-methylbenzyl-D-penicillamine dicyclohexylammonium salt

  • MDL number MFCD00235908
  • Cas Number : 198470-36-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-D-Pen(pMeBzl)-OH DCHA
Purity 98
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-s-i-4-methylbenzyl-d-penicillamine-dicyclohexylammonium-salt/1742?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-s-i-4-methylbenzyl-d-penicillamine-dicyclohexylammonium-salt/1742?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart