CH6371377736 Chem Impex International, Inc.

(3R)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid

  • MDL number MFCD00144038
  • Cas Number : 103733-65-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym D-Tic-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White to yellow solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-i-r-i-1-2-3-4-tetrahydroisoquinoline-3-carboxylic-acid/1747?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-i-r-i-1-2-3-4-tetrahydroisoquinoline-3-carboxylic-acid/1747?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart