CH6371377744 Chem Impex International, Inc.

Acetyl-L-alanine amide

  • MDL number MFCD00080821
  • Cas Number : 15962-47-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160-164 �C
Synonym Ac-L-Ala-NH2
Purity 99
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/acetyl-l-alanine-amide/1755?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/acetyl-l-alanine-amide/1755?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart