CH6371377749 Chem Impex International, Inc.

Acetyl-glycine N-hydroxysuccinimide ester

  • MDL number MFCD00153382
  • Cas Number : 24715-24-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99-105
Synonym Ac-Gly-OSu
Purity 98
Storage temp. Store at 0-8
Physical Form Off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/acetyl-glycine-i-n-i-hydroxysuccinimide-ester/1760?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/acetyl-glycine-i-n-i-hydroxysuccinimide-ester/1760?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart