CH6371377750 Chem Impex International, Inc.

Acetyl-glycine 4-nitrophenyl ester

  • MDL number MFCD00127756
  • Cas Number : 3304-61-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Ac-Gly-ONp
Purity 99
Storage temp. Store at 0-8
Physical Form White to slightly yellowish powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/acetyl-glycine-4-nitrophenyl-ester/1761?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/acetyl-glycine-4-nitrophenyl-ester/1761?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart