CH6371377756 Chem Impex International, Inc.

Acetyl-L-leucine amide

  • MDL number MFCD00026506
  • Cas Number : 28529-34-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129-135 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/acetyl-l-leucine-amide/1767?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/acetyl-l-leucine-amide/1767?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart