CH6371377767 Chem Impex International, Inc.

Acetyl-L-phenylalanine methyl ester

  • MDL number MFCD00066060
  • Cas Number : 3618-96-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 65-73
Synonym Ac-L-Phe-OMe
Purity 99
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/acetyl-l-phenylalanine-methyl-ester/1778?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/acetyl-l-phenylalanine-methyl-ester/1778?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart