CH6371377770 Chem Impex International, Inc.

Acetyl-L-proline amide

  • MDL number MFCD00037340
  • Cas Number : 16395-58-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148 - 150 °C
Purity 99
Storage temp. Store at RT
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/acetyl-l-proline-amide/1781?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/acetyl-l-proline-amide/1781?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart