CH6371377771 Chem Impex International, Inc.

Acetyl-L-proline methyl ester

  • MDL number MFCD00037338
  • Cas Number : 27460-51-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Ac-L-Pro-OMe
Purity 99
Storage temp. Store at RT
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/acetyl-l-proline-methyl-ester/1782?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/acetyl-l-proline-methyl-ester/1782?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart