CH6371377775 Chem Impex International, Inc.

Nalpha-Acetyl-L-tryptophan methyl ester

  • MDL number MFCD00022759
  • Cas Number : 2824-57-9
  • Molecular Weight : 260.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at RT
Physical Form White to off white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-acetyl-l-tryptophan-methyl-ester/1786?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-acetyl-l-tryptophan-methyl-ester/1786?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart