CH6371377777 Chem Impex International, Inc.

Acetyl-L-tyrosine amide

  • MDL number MFCD00002391
  • Cas Number : 1948-71-6
  • Molecular Weight : 222.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 224-228� C
Purity 99
Storage temp. Store at RT
Physical Form White powder
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/acetyl-l-tyrosine-amide/1788?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/acetyl-l-tyrosine-amide/1788?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart