CH6371377782 Chem Impex International, Inc.

Acetyl-L-valine amide

  • MDL number MFCD00080846
  • Cas Number : 37933-88-3
  • Molecular Weight : 158.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 234-242 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/acetyl-l-valine-amide/1793?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/acetyl-l-valine-amide/1793?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart