CH6371377791 Chem Impex International, Inc.

Z-N-methyl-L-isoleucine

  • MDL number MFCD00235876
  • Cas Number : 42417-66-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Z-N-Me-L-Ile-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-i-n-i-methyl-l-isoleucine/1803?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-i-n-i-methyl-l-isoleucine/1803?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart