CH6371377798 Chem Impex International, Inc.

Boc-N-methyl-L-norleucine (oil)

  • MDL number MFCD00153391
  • Cas Number : 117903-25-0
  • Molecular Weight : 245.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8°C
Physical Form Clear, yellow oil
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-n-i-methyl-l-norleucine-oil/1810?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-n-i-methyl-l-norleucine-oil/1810?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart