CH6371377802 Chem Impex International, Inc.

Na-Boc-Na-Methyl-L-phenylalanine dicyclohexylammonium salt

  • MDL number MFCD00070397
  • Cas Number : 40163-88-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 173-180� C
Synonym Boc-N-Me-L-Phe-OH DCHA
Purity 98
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-i-sup-a-i-sup-i-i-boc-i-n-i-sup-a-sup-methyl-l-phenylalanine-dicyclohexylammonium-salt/1814?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-i-sup-a-i-sup-i-i-boc-i-n-i-sup-a-sup-methyl-l-phenylalanine-dicyclohexylammonium-salt/1814?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart