CH6371377803 Chem Impex International, Inc.

Nalpha-Z-Nalpha-Methyl-L-phenylalanine dicyclohexylammonium salt

  • MDL number MFCD00235879
  • Cas Number : 2899-08-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117 - 119 °C
Purity 99
Storage temp. Store at 0 - 8 °C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-i-sup-a-i-sup-i-z-n-i-sup-a-sup-methyl-l-phenylalanine-dicyclohexylammonium-salt/1815?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-i-sup-a-i-sup-i-z-n-i-sup-a-sup-methyl-l-phenylalanine-dicyclohexylammonium-salt/1815?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart