CH6371377807 Chem Impex International, Inc.

Boc-N-methyl-O-benzyl-L-threonine cyclohexyl ammonium salt

  • MDL number MFCD06795764

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-N-Me-L-Thr(Bzl)-OH CHA
Purity 95
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-n-i-methyl-i-o-i-benzyl-l-threonine-cyclohexyl-ammonium-salt/1819?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-n-i-methyl-i-o-i-benzyl-l-threonine-cyclohexyl-ammonium-salt/1819?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart