CH6371377810 Chem Impex International, Inc.

Fmoc-N-methyl-O-benzyl-L-tyrosine

  • MDL number MFCD00270746
  • Cas Number : 428868-52-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 63-69 �C
Synonym Fmoc-N-Me-L-Tyr(Bzl)-OH
Purity 98
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-n-i-methyl-i-o-i-benzyl-l-tyrosine/1822?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-n-i-methyl-i-o-i-benzyl-l-tyrosine/1822?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart