CH6371377818 Chem Impex International, Inc.

Boc-L-aspartic acid b-9-fluorenylmethyl ester

  • MDL number MFCD00076916
  • Cas Number : 117014-32-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-L-Asp(OFm)-OH
Purity 98
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-l-aspartic-acid-ß-9-fluorenylmethyl-ester/1830?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-l-aspartic-acid-ß-9-fluorenylmethyl-ester/1830?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart