CH6371377830 Chem Impex International, Inc.

Boc-L-aspartic acid a-cyclohexyl ester dicyclohexylammonium salt

  • MDL number MFCD00076914
  • Cas Number : 200283-00-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 151-160� C
Synonym Boc-L-Asp-OcHex DCHA
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-l-aspartic-acid-a-cyclohexyl-ester-dicyclohexylammonium-salt/1842?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-l-aspartic-acid-a-cyclohexyl-ester-dicyclohexylammonium-salt/1842?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart