CH6371377832 Chem Impex International, Inc.

Boc-L-glutamic acid a-cyclohexyl ester

  • MDL number MFCD00076928
  • Cas Number : 137506-13-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 89-93 �C
Synonym Boc-L-Glu-OcHex
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-l-glutamic-acid-alpha-cyclohexyl-ester/1844?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-l-glutamic-acid-alpha-cyclohexyl-ester/1844?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart