CH6371377834 Chem Impex International, Inc.

Boc-His(3-Me)-OH

  • MDL number MFCD00076945
  • Cas Number : 61070-22-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8°C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-his-3-me-oh/1847?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-his-3-me-oh/1847?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart