CH6371377836 Chem Impex International, Inc.

Boc-S-benzyl-L-homocysteine

  • MDL number MFCD00080371
  • Cas Number : 16947-99-2
  • Molecular Weight : 325.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 101-107° C
Purity 98
Storage temp. Store at 0-8°C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-s-i-benzyl-l-homocysteine/1849?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-s-i-benzyl-l-homocysteine/1849?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart