CH6371377837 Chem Impex International, Inc.

Boc-S-4-methylbenzyl-L-homocysteine

  • MDL number MFCD00080372
  • Cas Number : 201419-15-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 84 - 89 °C
Purity 99
Storage temp. Store at 0 - 8 °C
Physical Form White product
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-s-i-4-methylbenzyl-l-homocysteine/1850?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-s-i-4-methylbenzyl-l-homocysteine/1850?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart