CH6371377840 Chem Impex International, Inc.

Na-Boc-Ne-dimethyl-L-lysine

  • MDL number MFCD00076965
  • Cas Number : 65671-53-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-L-Lys(Me)2-OH
Purity 98
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-boc-i-n-i-sup-e-sup-dimethyl-l-lysine/1853?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-boc-i-n-i-sup-e-sup-dimethyl-l-lysine/1853?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart