CH6371377841 Chem Impex International, Inc.

Nalpha-Boc-Nepsilon-isopropyl-Nepsilon-Z-L-lysine dicyclohexylammonium salt

  • MDL number MFCD00076967

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 59-69 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-boc-i-n-i-sup-epsilon-sup-isopropyl-i-n-i-sup-epsilon-sup-z-l-lysine-dicyclohexylammonium-salt/1854?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-boc-i-n-i-sup-epsilon-sup-isopropyl-i-n-i-sup-epsilon-sup-z-l-lysine-dicyclohexylammonium-salt/1854?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart