CH6371377843 Chem Impex International, Inc.

Na-Boc-Ne-nicotinoyl-L-lysine

  • MDL number MFCD00076969
  • Cas Number : 14609-04-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 131-138
Synonym Boc-L-Lys(nicotinoyl)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-boc-i-n-i-sup-e-sup-nicotinoyl-l-lysine/1856?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-boc-i-n-i-sup-e-sup-nicotinoyl-l-lysine/1856?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart