CH6371377845 Chem Impex International, Inc.

Na-Fmoc-Ne-nicotinoyl-D-lysine

  • MDL number MFCD00080277
  • Cas Number : 252049-12-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point > 200 �C
Synonym Fmoc-Nepsilon-nicotinoyl-D-lysine
Purity 98
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-fmoc-i-n-i-sup-e-sup-nicotinoyl-d-lysine/1858?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-fmoc-i-n-i-sup-e-sup-nicotinoyl-d-lysine/1858?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart