CH6371377860 Chem Impex International, Inc.

Boc-O-benzyl-3,5-dibromo-L-tyrosine

  • MDL number MFCD00076992
  • Cas Number : 218769-47-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 143-149
Synonym Boc-3,5-dibromo-L-Tyr(OBzl)-OH
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-o-i-benzyl-3-5-dibromo-l-tyrosine/1874?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-o-i-benzyl-3-5-dibromo-l-tyrosine/1874?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart