CH6371377862 Chem Impex International, Inc.

Boc-N-methyl-O-2,6-dichlorobenzyl-L-tyrosine

  • MDL number MFCD00038768
  • Cas Number : 57817-43-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 121-125° C
Purity 100
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-n-i-methyl-i-o-i-2-6-dichlorobenzyl-l-tyrosine/1876?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-n-i-methyl-i-o-i-2-6-dichlorobenzyl-l-tyrosine/1876?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart