CH6371377870 Chem Impex International, Inc.

3-(1-Naphthyl)-L-alanine

  • MDL number MFCD00066086
  • Cas Number : 55516-54-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Ala(1-naphthyl)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-1-naphthyl-l-alanine/1937?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-1-naphthyl-l-alanine/1937?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart