CH6371377910 Chem Impex International, Inc.

Boc-O-benzyl-D-threonine

  • MDL number MFCD00038247
  • Cas Number : 69355-99-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110-117°C
Purity 99
Storage temp. Store at 0-8°C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-o-i-benzyl-d-threonine/1982?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-o-i-benzyl-d-threonine/1982?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart